Certified verification thermometers
Supplier: Amarell
AMARA303421EA 0 EUR
AMARA303421 AMARA303541 AMARA303665 AMARA303621 AMARA303401 AMARA304631 AMARA303521 AMARA303645 AMARA303469 AMARA303655 AMARA303501 AMARA303611 AMARA303191 AMARA303171 AMARA303291 AMARA304251 AMARA303051 AMARA304441 AMARA303571 AMARA304771 AMARA303031 AMARA303581 AMARA303881 AMARA303331
Certified verification thermometers
These glass thermometers with ambitious approval have a validity of the verification of 15 years.
  • Solid stem
  • Durable pigment
  • Top finish round, in transparent
Mercury, toluen (AMARA304531)
Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to be used for tests as per standards which require the usage of mercury thermometers.
Certifications: ASTM standards
Packaging: Supplied in a square plastic case.