Occupational shoes, clogs, Easy, 7622/7623
Supplier: Abeba
Easy
ABEB7622/35EA 61.56 EUR
ABEB7622/35 ABEB7623/36 ABEB7623/37 ABEB7623/38 ABEB7622/38 ABEB7623/39 ABEB7623/43 ABEB7622/43 ABEB7622/44 ABEB7623/35 ABEB7622/39 ABEB7623/40 ABEB7623/41 ABEB7623/42
Occupational shoes, clogs, Easy, 7622/7623
Shoes and Boots
Anti-static clog made from smooth leather with a non foldable, adjustable heel strap. The TPU sole is resistant to fuel, oil and animal fats.
  • Replaceable acc Wave insole; orthopaedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • TPU sole with high slip resistance
Colour: White/blue or white/red
Certifications: DGUV 112-191; EN ISO 20347:2012; OB, FO, E, A, SRC