Anti-vibration pads
Supplier: Eppendorf
EPPE5181301.009EA 0 EUR
EPPE5181301.009 EPPE5181309.000 EPPE5181307.007 EPPE5181303.001 EPPE5181305.004
Anti-vibration pads
Surface Protectors Bench Protectors
The anti-vibration pads are specifically designed to effectively protect micromanipulation systems against extreme vibrations. The pads are directly positioned under the base points of the microscope. The various pads are optimised for specific load ranges to guarantee perfect results.
  • Effectively protects micromanipulation workstations from extreme external vibrations
  • Five different sizes available
  • Different sizes combinable for optimal adjustment