Hand cleansing pastes, Kresto® Kolor ULTRA/Kresto® colour
Supplier: SC Johnson
EVONKKU250MLEA 75 EUR
EVONKKU250ML
Hand cleansing pastes, Kresto® Kolor ULTRA/Kresto® colour
Skincare Soaps and Skin Cleansers
Special hand cleanser paste for the removal of stubborn dyes with a very powerful cleansing action. For contamination which are reducible within the alkaline range, like toner, aniline dyes, azo dyes, printing inks, copying inks, fruit and vegetable colourants.
  • Contains active agent for easy removal of inks and dyes from the skin
  • Astopon® natural scrubbing agent (refined walnut shell powder) gently removes contaminants without damaging the skin
  • As the formulation is concentrated, only a small amount is required to remove contaminants
  • Solvent-free, colourant-free, preservative-free, perfumed
Ordering information: Soft bottle may only be used with Stoko® Vario® mat dispensers.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904