Thermometer for distillation processes
Supplier: Behr
BEHR6237293EA 0 EUR
BEHR6237293
Thermometer for distillation processes
This replaceable thermometer is mercury-free.
This replaceable thermometer is mercury-free.
Red filling.
Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to be used for tests as per standards which require the usage of mercury thermometers.