Ethidium bromide 0,5 mg/ml in aqueous solution, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
SIALE1385-5MLEA 24.8 EUR
SIALE1385-5ML
Ethidium bromide 0,5 mg/ml in aqueous solution, Sigma-Aldrich®
Ethidium bromide
Formula: C₂₁H₂₀BrN₃
MW: 394,31 g/mol
MDL Number: MFCD00011724
EINECS: 214-984-6