Homogeniser, T 10 standard ULTRA-TURRAX® PCR kit
Supplier: IKA
ULTRA-TURRAX®
431-0174EA 0 EUR
431-0174 431-0173
Homogeniser, T 10 standard ULTRA-TURRAX® PCR kit
Homogenisers Homogenisers Handheld
Handheld or stand model, volume range 0,5 to 100 ml. A wide speed range allows homogeniser to work at high circumferential speeds even with small rotor diameters.
  • Quick-release coupling makes changing the dispersing elements easy
  • Excellent speed stability with various media
  • Disposable plastic dispersing elements are available in two sizes. These are ideal for PCR analysis
Certifications: IP protection class according to DIN EN 60529: IP 30; operating temperature range 5 to 40 °C
Delivery information: Package includes the ULTRA-TURRAX® unit, clamp R 200, storage case with dispersing elements S10D-7G-KS-65 and S10D-7G-KS-110 and a set of 12 dispersing elements S10D-7G-KS-65.