Ice baths
Supplier: NeoLab
478-1255EA 69 EUR
NEOL1-6031
Ice baths
Extremely good isolated ice bath made of double-walled polystyrene. The hollow is foamed in the inside for a better temperature isolation. Due to it a large insulation is achieved.
  • Ice formations are remaining very long
  • Fill with ice pieces or brine for cooling
  • Temperature resistance: –40 to +90 °C