Electromantle
Supplier: Electrothermal
Electrothermal
ELETCMUT1000/CEEA 919 EUR
ELETCMUT1000/CE
Electromantle
Heaters Heating Mantles
The CMUT electromantle is used in wet chemistry to heat liquids in round bottom flasks. The metal case is designed to remain cool-to-the-touch when in operation. For added safety, it has a grounded stainless steel earth screen covering the element and is double fused.
  • Element temperatures up to 450 °C
  • Built-in energy regulator
  • Indicator lamps for power and heater operation
  • Replaceable insulated heater cartridge
Rugged and easy to clean powder coated aluminium outer casing. Non-skid feet and support rod bracket (but not including support rods). Flexible coiled heating elements absorb shock, minimising the risk of flask breakage. Heating element is attached to a rigid thermal insulating cartridge to ensure maximum heat transfer and support.