Vascular endothelial growth factor (VEGF), Corning®
Supplier: Corning
BDAA354107EA 0 EUR
BDAA354107
Vascular endothelial growth factor (VEGF), Corning®
VEGF is a member of the PDGF family with endothelial cell-specific activities (e.g., angiogenic and mitogenic factor).
Human recombinant, a dimeric glycoprotein composed of two A and / or B chains and is the principal mitogen in serum for mesenchymal cells, has endothelial cell-specific activities.
Tested for ability to promote proliferation of HUVEC. Filtered (0,2 µm membrane) and tested and found negative for the presence of bacteria, fungi, and mycoplasma. Tested for endotoxin (LAL assay).