You Searched For: Bossheads
44  results were found
SearchResultCount:"44"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Bossheads made from different materials for universal usage.
Supplier: VWR Collection
Description: For assembling 8, 12, 13, 16 and 21 mm diameter rods.
Supplier: VWR Collection
Description: For assembling 12, 13 and 16 mm diameter rods.
Catalog Number: (241-0113)
Supplier: VWR Collection
Description: Bosshead made of laboral aluminium alloy for connecting two retort rods at any angle.
UOM: 1 * 2 items
Catalog Number: (241-2550)
Supplier: Lenz Laborglas GmbH & CO.KG
Description: Bosshead made from tempered cast iron. For attaching reaction vessel clamps to support stands.
UOM: 1 * 1 items
Catalog Number: (241-0371)
Supplier: USBECK Laborgeräte
Description: Zinc die-casting, nickel plated.
UOM: 1 * 1 items
Supplier: USBECK Laborgeräte
Description: Bossheads made of zinc die-cast, either natural, powder coated or nickel plated.
Supplier: USBECK Laborgeräte
Description: For stand assembly.
Supplier: USBECK Laborgeräte
Description: Bosshead, Hook connector, Steel, powder coated, For rod Ø: 13 mm
Catalog Number: (USBE2240)
Supplier: USBECK Laborgeräte
Description: Bosshead made of powder-coated zinc die-cast, with nickel plated brass screws.
UOM: 1 * 1 items
Catalog Number: (241-0370)
Supplier: USBECK Laborgeräte
Description: Double bosshead made from powder-coated aluminium alloy.
UOM: 1 * 1 items
Supplier: USBECK Laborgeräte
Description: Vierkantmuffe, Material: Aluminium, für Stativstäbe mit Dmr: bis 13 mm, mit Lappenschrauben 1 * 1 items
New Product
Catalog Number: (241-0250)
Supplier: USBECK Laborgeräte
Description: Bossheads made of powder-coated zinc-die casting.
UOM: 1 * 1 items
Supplier: BOCHEM
Description: Cross type double bossheads, either made of stainless steel or malleable cast iron, chromium plated.
Supplier: BOCHEM
Description: Bosshead with heart screw, Spanning width: 13 mm
Supplier: BOCHEM
Description: Three angled bossheads made of duraluminium (aluminium alloy).
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 44
no targeter for Bottom