Next generation sequencing, PerfeCTa® NGS Quantification Kit
Supplier: Quantabio
733-2300EA 680 EUR
QUNT95154-500 QUNT95155-500 QUNT95156-500
Next generation sequencing, PerfeCTa® NGS Quantification Kit
Accurate quantification of the number of amplifiable library molecules is the most critical step in the Next Generation Sequencing (NGS) workflow in obtaining high quality read data with next generation sequencing technologies. PerfeCta® NGS Quantification kits use Real-Time PCR with prediluted standards for the convenient and accurate quantification of NGS libraries for the Illumina® NGS platform.
  • Accurate and sensitive method for NGS library quantification
  • Stabilised, prediluted standards for convenient use
  • Consistency across a broad range of samples
  • Sensitivity of detection to quantify low concentration libraries
  • Quantification for Illumina®