These waterproof pocket meters provide direct measurements of different ions in liquid, solid, viscous or powder samples using a unique flat sensor with an ion selective membrane.
- Auto calibration function recognises standards and is user-selectable for either 1- or 2-point calibrations
- Auto range change, temperature compensation
- Auto hold, automatic power off after 30 minutes
- Easy to use, very convenient for quick on-site testing
- Use of sampling sheets enables measurements in micro samples as small as 0,05 ml
The user only needs to drip a few drops of the sample on to the sensor in order to get quick results − there's no need to dilute or prepare the samples beforehand as these reagent-free meters have a wide measurement range. Samples can be as small as 0,3 ml. These meters are suitable for research and industrial applications in diverse fields, such as agriculture and food testing.
Delivery information: Each meter is supplied with two batteries, pipette, instruction manual, quick manual, storage case, two bottles of standard solutions (150 and 2000 ppm), 14 ml each, and five pieces of sampling sheet B.