Disinfectant wipes for surface cleaning, Bacillol® Wipes
Supplier: Bode
Bacillol® Wipes
BODE976350EA 0 EUR
BODE976350
Disinfectant wipes for surface cleaning, Bacillol® Wipes
Wipes Disposable Wipes and Towels Disinfectant Wipes
Bacillol® Wipes are alcohol-based quick disinfection wipes for convenient, daily disinfection of alcohol resistant surfaces, such as work surfaces in medical practices, labs and clinics, and medical equipment.
  • Wipes can be easily folded, allowing for proper moistening of smaller and angled surfaces and recesses
  • Tear-resistant and absorbent fleece
  • Easy dispensing from the X-Wipes dispensing system
  • Broad range of applications and material compatibility
  • Bacillol® AF saturation fluid guarantees effective moistening
It is especially advantageous to use the wipes in combination with the X-Wipes dispensing system, in a work environment where transferring alcohol-based disinfectants is not permitted (e.g. for fire or work safety reasons).
Microbiological effectiveness: bactericidal, yeasticidal, fungicidal, tuberculocidal, mycobactericidal, limited virucidal action (incl. HBV, HIV, HCV), MNV, adeno-, polyoma- and rotaviruses.
VAH, Robert Koch Institute (RKI) list of tested and recognised disinfectants and methods.
Packaging: 90 wipes per stand-up bag. This can be inserted in the Bacillol® Wipes dispenser or in an X-Wipes dispenser.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904