Flame retardant rain overalls, Houston 6969
Supplier: Sioen
SIOE6969/H45/1XLEA 146 EUR
SIOE6969/H45/1XL SIOE6969/H45/2XL SIOE6969/H45/M SIOE6969/H45/L SIOE6969/H45/3XL SIOE6969/H45/S
Flame retardant rain overalls, Houston 6969
These unlined overalls are made of Flexothane® Flame fabric consisting of 100% FR polyester knitting with FR PU coating. Garments feature high frequency welded seams.
  • Extended durability
  • Fixed hood with peak, zip closure under double flap with press studs
  • Raglan sleeves, sleeve and ankle narrowing by press studs
  • Elasticated windcuffs in sleeves
  • Elasticated waist in the back
Weight: 190 g/m²
Certifications: EN ISO 14116 (index 1), EN 343 (3-1)