Microinjection capillaries, Femtotip® II
Supplier: Eppendorf
631-0237EA 137 EUR
631-0237
Microinjection capillaries, Femtotip® II
Microinjection Capillaries
Microinjection capillary for reproducible injection of adherent and suspension cells.
  • Defined opening with 0,5 µm inner diameter and 0,7 µm outer diameter (±0,1 µm)
  • Screw-thread for rapid mounting
  • Precise length accuracy
  • 100% individual testing of every Femtotip® II using a flow parameter