Microinjection capillaries, VacuTip I
Supplier: Eppendorf
VacuTip
EPPE5195000.036EA 477 EUR
EPPE5195000.036
Microinjection capillaries, VacuTip I
Microinjection Capillaries
Glass microcapillary for holding suspension cells (oocytes or blastocysts).
  • 15 μm inner diameter, 100 µm outer diameter
  • 35° tip angle
  • Flange length 1 mm
  • Smooth rounded tip end for gentle holding
  • Sterilised by gamma irradiation
  • Non cytotoxic (as proven by mouse embryo assay)
  • Capillary safe for optimum protection
Caution: For research use only.