VWR® kuroGEL Maxi Plus 25 Horizontal Electrophoresis System
Supplier: Avantor
700-0154EA 0 EUR
VWRI700-0154
VWR® kuroGEL Maxi Plus 25 Horizontal Electrophoresis System
The kuroGEL Maxi-Plus 25 Horizontal gel electrophoresis unit is ideal for high throughput analysis of nucleic acids. Large format, 25×30 cm, gel tray is ideal for high throughput techniques, such as HLA typing and screening PCR products.
  • Multi channel pipette compatible combs, with a maximum 52-sample throughput, allow samples to be loaded quickly and easily from 96- and 384-well thermal cycler blocks
  • Buffer recirculation ports can be connected to a peristaltic pump for buffer recirculation during electrophoresis to maintain buffer pH and prevent ionic gradient formation
  • Coloured loading strips, for easy well detection when loading
  • UV-transparent acrylic casting tray allows the user to handle the gel on the transilluminator with minimum risk of exposure to hazardous ethidium bromide
  • Side handles for safe and easy transportation around the laboratory
Gel tank supplied with removable casting tray; 2×1,0 mm thick, 26-sample combs; coloured loading strips and buffer recirculation ports.
Combs for kuroGEL Maxi Plus 25
Combs are colour coded and height adjustable, offering complete control over loading volume and well depth to a maximum 624-sample throughput. Twelve comb positions at 2 cm intervals along the tray for rapid, high throughput separation of multiple samples.
Comb slots: 12; comb thickness: 1,0, 1,5 or 2,0 mm; comb throughput: 26 to 52 samples.
Accessories for kuroGEL Maxi Plus 25
Casting options: Casting gates with integral silicone seals effectively seal the tray without the need for tape, provided the silicone gasket faces outwards. 25 cm long Super-Seals offer total versatility in casting, allowing the gel length to be tailored to each user's personal requirements.
Coloured loading strips (optional) aid visualisation during gel loading. Available in three different colours for easy visualisation of samples mixed with dyes such as bromophenol and xylene cyanol. Each colour strip has an adhesive backing allowing it to be easily affixed and removed with each application.
Gel scoops (optional) are ideal for safe and easy transfer of gels from staining or destaining tanks to a transilluminator or similar surface.
*Multi channel pipette compatible