Medium pressure 6-way selection valves, Upchurch Scientific®
Supplier: IDEX Health & Science
Upchurch Scientific®
560-0166EA 815 EUR
560-0166 560-0167
Medium pressure 6-way selection valves, Upchurch Scientific®
Valves Selection Valves
The 6-way medium pressure selection valves have a common inlet (or outlet) port connected to six different ports. If the application does not require all six outer ports, seal the extra port(s) with any flat-bottom ¹/₄ - 28 plug.
  • Biocompatible and inert
  • Chemically resistant Vespel® and PTFE resin-wetted surfaces
  • Pressure rated to 1000 psi (69 bar)
  • Excellent choice for mobile phase selection
Ordering information: Each valve includes six ¹/₄ - 28 flangeless fittings for ¹/₁₆" or ¹/₈" ext. Ø tubing.