Pipette washer sets, Azlon®
Supplier: SciLabware
AZLORDR306EA 0 EUR
AZLORDR306 AZLORDR302 AZLORDR304
Pipette washer sets, Azlon®
Pipette rinser is an automatic rinsing system ensuring rapid washing with high-quality results.
  • Automatic rinsing system ensuring rapid washing with high-quality results
  • Suitable for use with pipette basket and pre-cleaning jar
  • Automatically rinses contents when connected to a tap and drain
  • Rinses pipettes, burettes and other labware up to 600 mm in length
Pipette pre-cleaning jar: For pre-cleaning pipettes in cleaning solution prior to rinsing in the pipette rinser.
Pipette basket: For dipping pipettes into the pre-cleaning jar or pipette rinser. Suitable for transferring pipettes safely.