Tubing, LDPE, fine-bore
Supplier: Smiths Medical
SMME800/100/280EA 82.4 EUR
SMME800/100/680100 SMME800/100/605100 SMME800/100/280 SMME800/100/500100 SMME800/100/540100
Tubing, LDPE, fine-bore
Tubing Transfer Tubing
PE, translucent.
  • Lightweight and flexible
  • High chemical resistance
  • Smooth bore, easily cleaned
This tubing has surgical non-toxic quality, minimises trauma when used in vivo. Suitable for sheathing delicate glass fibres in telecommunications.