You Searched For: Hygiene Masks
9  results were found
SearchResultCount:"9"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Face masks made from 3 ply, non woven PP featuring a nose bridge and ear loops or ties.
Sale
Catalog Number: (76470-110)
Supplier: VWR Collection
Description: VWR® made in USA sterile masks are an ideal option for single-use needs.
UOM: 1 * 200 items
Catalog Number: (114-9101)
Supplier: HUBEI ORIENT INT'L TRADING COR
Description: Hygiene masks made from 3-ply construction of PP and Meltblown PP.
UOM: 1 * 50 items
Catalog Number: (HOIT115688)
Supplier: HUBEI ORIENT INT'L TRADING COR
Description: Face mask with comfortable elastic ear loops for easy donning.
UOM: 1 * 10.000 items
Catalog Number: (113-0713)
Supplier: VWR Collection
Description: This face mask, with nose bridge, is made from two ply, non woven PP.
UOM: 1 * 50 items
Supplier: DESCO
Description: These facemasks constructed of a lightweight dissipative material which is made from texturised polyester and a minimum of 9% carbon nylon monofilimen...
Catalog Number: (113-9935)
Supplier: ALSICO HIGHTECH NV
Description: Reusable, 2-ply face masks are not designed for medical use. Masks are not tested on the permeability of viruses and bacteria.
UOM: 1 * 1 items
Supplier: DELTALYO & VALMY
Description: The FFP3 cup-shaped mask is both reassuring and light weight. The respiratory tract is shrouded in 10 g of comfort.
Catalog Number: (113-9934)
Supplier: RENTEX COM D.O.O.
Description: Made from 100% one layer cotton, this fine twill mask can be washed on higher temperatures for repeated use.
UOM: 1 * 250 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 9 of 9
no targeter for Bottom