Calorimeters, C 6000 global standards and isoperibol packages
Supplier: IKA
IKAA8804700EA 0 EUR
622-0036 622-0037 IKAA8804700 IKAA8804800 IKAC10001047 622-0038 IKAC10001048 622-0039 IKAC10001049 662-2321 662-2325 662-2324 662-2323 662-2322 622-0042 662-2328 IKAA10001049 662-2327 IKAA10001048 662-2326 IKAA10001047 IKAA10001046 IKAA8804300 IKAA8804400
Calorimeters, C 6000 global standards and isoperibol packages
Calorimeters
The C 6000 global standards and C 6000 isoperibol calorimeters follow the approved C 5000 and C 2000 calorimeter models.
  • Convenient touch screen operation
  • Software provides control chart view and correction calculation
  • RFID technology used for decomposition vessel identification
  • SD card slot, Ethernet interface for additional data management
  • Decomposition vessel with spherical top for faster heat transfer results in shorter experimental times
  • Easy bomb preparation due to 'turned around' crucible holder technology
The C 6000 global standards offers a fast dynamic method, the classical adiabatic as well as isoperibol measurement modes. The C 6000 isoperibol offers the same advantages and features, with the exception of the adiabatic measurement mode. Each apparatus can be operated through a user panel and with the software Calwin C 6040. This software opens up further features in data handling with Microsoft SQL, LIMS and correction calculations that follow many calorimeter standard.