Sponges, MegaTex™
Supplier: Micronova
MegaTex®
129-0638EA 16.6 EUR
129-0638
Sponges, MegaTex™
Wipes Sponges
These sponges are made of MegaTex® fabric (non woven polyamide fabric) over a urethane foam interior. Ideal for use on workstations, laminar flow benches, walls and windows.
  • Textured thermo-bonded surface
  • Hold up to strong disinfectants and steam sterilisation
  • Ideal for cleaning heavy soils, structured floors and other rough surfaces