Sponge, NovaCel™
Supplier: Micronova
NovaCel™
129-0639EA 520 EUR
129-0639
Sponge, NovaCel™
Wipes Sponges
These cleaning sponges are made of PVA (polyvinyl alcohol) and absorb 25 times their weight. Their ultra-clean quality and broad chemical resistance allow them to be used for lapping and polishing in wafer manufacture as well as general wiping applications.
  • Withstand strong disinfectants and steam sterilisation
  • Firm to the touch in their dry state, but once hydrated the material becomes extremely soft and pliant
  • Launderable and autoclavable