E.Z.N.A.® Plant RNA Midi Kit
Supplier: Omega Bio-Tek
R6628-00EA 29.7 EUR
OMEGR6628-00
E.Z.N.A.® Plant RNA Midi Kit
The E.Z.N.A.® Plant RNA Midiprep Kit provides a convenient and rapid method for the isolation of total RNA from a variety of plant samples. These kits are designed for preparation of RNA from large-scale plant samples and include shredding/homogenizing columns to efficiently remove cell debris and simultaneously homogenise the lysate. In combination with HiBind® RNA spin columns, this permits purification of high quality RNA from as much as 500 mg plant tissue in less than 60 minutes. Purified RNA is suitable for RT-PCR, Northern analysis, differential display, and Poly A+ RNA selection.
  • RNA isolation under 60 minutes
  • Optimised buffer system ensure the reproducible results
  • No organic extractions
  • Purified RNA suitable for most downstream applications