E.Z.N.A.® PF Micro RNA Kit
Supplier: Omega Bio-Tek
R7036-00EA 41 EUR
OMEGR7036-00 OMEGR7036-01
E.Z.N.A.® PF Micro RNA Kit
The E.Z.N.A.® PF Micro RNA Kit combines the reversible binding properties of HiBind® matrix, a silica-based material, with the unique lysis and binding procedure to extract micro and large (>200 nt) RNA from a wide variety of starting materials. A specially formulated high salt lysis and binding buffer system allows more than 100 µg of RNA to bind to the matrix.
  • No organic extractions
  • Efficient isolation of small RNA containing total RNA
  • Enrich for small RNA <200 nt to increase sensitivity in downstream analysis
  • Simple 30 minutes procedure
  • Compatible with virtually all cell and tissue type