You Searched For: Salinity Meters
7  results were found
SearchResultCount:"7"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: ATAGO
Description: These salt meters utilise the electric conductivity method and require no reagent solutions. They feature a funnel-shaped sensor section which measure...
Catalog Number: (ATAG4211)
Supplier: ATAGO
Description: This digital salt meter features a funnel-shaped sensor section (conductivity), which measures the salt % of a sample (dilute solution of soy sauce, k...
UOM: 1 * 1 items
Catalog Number: (LOVI194804-16)
Supplier: Lovibond
Description: The SD 90 salt meter is a compact, easy to use handheld meter for the measurement of salinity.
UOM: 1 * 1 items
Catalog Number: (EUTEECOSALTTEST)
Supplier: EUTECH
Description: This waterproof and accurate tester with stainless pin sensor is ideal for basic salinity measurements in aquaculture, spas, pools, simple lab work an...
UOM: 1 * 1 items
Catalog Number: (EBRO1340-5211)
Supplier: EBRO
Description: The SSX 210 salt meter is used to measure the salt content in semi-solid food products, such as meat, cold cuts, cheese, salads etc. To achieve this, ...
UOM: 1 * 1 items
Catalog Number: (HORI3200689158)
Supplier: HORIBA
Description: This meter directly measures sodium ion concentration in 0,3 ml sample (or 0,05 ml with sampling sheet B) and converts it into salinity or NaCl salt c...
UOM: 1 * 1 items
Catalog Number: (HORI3999960128)
Supplier: HORIBA
Description: This waterproof meter features a unique flat sensor that enables quick, accurate and direct measurements of micro samples as small as 0,12 ml. The sen...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 7 of 7
no targeter for Bottom