Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (43)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
digital-thermometers-with-probe
SearchPresentationType-VERTICAL
SearchResultCount:"41"
  List View Grid View Easy View (new)
Sort by:
Supplier:  VWR Collection
Description:   These digital stem thermometers are suitable for field studies, wet lab applications, wash-down areas, food testing and tanks.
Supplier:  VWR Collection
Description:   This 'flip to open' thermometer has a probe that can be positioned at any angle.
UOM:  1 * 1 items
Supplier:  VWR Collection
Description:   These highly accurate logging thermometers are designed to monitor temperatures in refrigerators, freezers, water baths, heating blocks and incubators...
Supplier:  VWR Collection
Description:   Data logging thermometers with user-defined timing intervals. Ideal for monitoring ultra-low temperature freezers.
Supplier:  VWR Collection
Description:   These high accuracy digital thermometers, with chemical resistant probes, monitor the temperature of pharmaceuticals, vaccines, chemicals, blood and b...
Supplier:  VWR Collection
Description:   This waterproof and shockproof thermometer is a safe replacement for any hazardous mercury instrument. The probe is resistant to most chemicals.
Supplier:  VWR Collection
Description:   Safe thermometer for soil, cuvettes, test tubes, food products, incubators, petroleum hydrocarbons and virtually all lab tests. Readings are updated e...
Catalog Number: (620-1975)
Supplier:  VWR Collection
Description:   Digital thermometer with extra long waterproof stainless steel 500 mm probe with cable (2,5 m).
UOM:  1 * 1 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 41
  1  2  3  4  5  6  Next