You Searched For: Analogue Thermometers
118  results were found
SearchResultCount:"118"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (620-2146)
Supplier: VWR Collection
Description: This thermometer, with analogue dial, indicates freezer, refrigerator, or spoilage temperatures.
UOM: 1 * 1 items
Supplier: VWR Collection
Description: Contains red organic liquid against white back glass, except for 620-0891 which has yellow back glass.
Supplier: VWR Collection
Description: Durac Plus precision thermometers are made with permanently fused markings and fractional degrees to provide precision and ensure accuracy.
Supplier: VWR Collection
Description: These glass thermometers can be used to take readings in freezers, refrigerators, incubators, ovens and blood banks.
Catalog Number: (620-0876)
Supplier: VWR Collection
Description: Dual-scale thermometer (°C/°F) with completely enclosed bulb for accurate readings. Easy to read on white-backed glass.
UOM: 1 * 1 items
Supplier: VWR Collection
Description: These thermometers are easy to read quickly and accurately due to the high contrast black numbering against a yellow background. Ideal for use in chem...
Supplier: VWR Collection
Description: These thermometers are easy to read quickly and accurately due to the high contrast black numbering against a yellow background. The numbers are print...
Supplier: VWR Collection
Description: These mercury-free thermometers are individually calibrated and useful for verifying sample temperatures. They are designed for applications in freeze...
Supplier: VWR Collection
Description: These oven thermometers contain red organic liquid against white back glass.
Supplier: VWR Collection
Description: These thermometers are suitable for applications with freezers, refrigerators, incubators, ovens and blood banks. The thermometer is placed in a bottl...
Supplier: VWR Collection
Description: These thermometers meet all requirements for physical dimensions and accuracy.
Catalog Number: (620-0920)
Supplier: VWR Collection
Description: This maximum/minimum thermometer displays the current temperature and registers the maximum and minimum temperatures attained since the last reset. It...
UOM: 1 * 1 items
Supplier: VWR Collection
Description: Each easy to read thermometer includes an individualised thermometer calibration report stating corrections to one-tenth of the smallest division, uni...
Supplier: VWR Collection
Description: These liquid-in-glass thermometers feature blue organic liquid fill against lead-free white back glass. They are ideal for use in chemical laboratorie...
Catalog Number: (620-0892)
Supplier: VWR Collection
Description: This thermometer has been designed to indicate critical temperature range. The critical temperature range from 4 to 6 °C is marked with arrows.
UOM: 1 * 1 items
Catalog Number: (620-0801)
Supplier: VWR Collection
Description: This high precision, EnviroKleen™ certified thermometer is accurate to ±1,0 °C and features green, non toxic, enviro safe liquid against lead-free, wh...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 118
no targeter for Bottom