Precast gels, GeneGel Excel 12,5/24 Kit
Supplier: Cytiva
17-6000-14EA 0 EUR
GEHE17-6000-14
Precast gels, GeneGel Excel 12,5/24 Kit
GeneGel Excel 12,5/24 kit is a ready to run gel and electrode system that highlights the powerful bufferless setup and handling ease of flatbed electrophoresis systems.
  • 24 sample wells accommodate up to 6 μl sample
  • Separation zone, T = 12,5%, C = 2%
Designed to service the broadest range of standard DNA applications.