Cryostat, CM 1520
Supplier: Leica
720-2268EA 0 EUR
720-2268
Cryostat, CM 1520
Microtomes and Microtome Assemblies Microtomes
The CM1520 offers exceptional reliability and low maintenance thanks to a fully encapsulated microtome, a removable object head for easy cleaning and a field-proven microtome design. Extra calm fan, substantially reducing the noise output of the working unit.
  • Precise control and exceptional reliability
  • Advanced ergonomics
  • Automatic hot gas defrost, maximum freezing –35 °C
High precision microtome with stepper motor feed-in and accurate specimen centering with zero-position.
Cutting stroke: 59 mm
Max specimen size: 55×55 mm or 55×80 mm