Ammonium peroxodisulphate (APS) 150 mg, tablets, Biotechnology Grade
Supplier: VWR Chemicals
Synonyms: Ammonium persulphate
K833-100TABSEA 103 EUR
K833-100TABS
Ammonium peroxodisulphate (APS) 150 mg, tablets, Biotechnology Grade
Ammonium peroxodisulphate (APS)
Ammonium persulphate forms free radicals when dissolved in water, which initiate polymerisation of acrylamide solutions.
  • Convenient tablet, easily prepared without weighing
  • Fast and easy polymerisation of acrylamide mixtures
  • Odour-free and non flammable
  • Non hazardous shipping
Due to the instability of APS in water, a working solution must be prepared daily for optimum performance. A 10% APS solution can be easily be prepared, without weighing, by adding one 150 mg APS tablet to 1,5 ml of water.
Formula: (NH₄)₂S₂O₈
MW: 228,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003390
CAS Number: 7727-54-0
Order Now
Specification Test Results
Dissolution Time (1 tablet/1mL Water) <= 4 minutes
Polymerization Time <= 30 minutes
Solubility (10%, Water) PASS P/F
Tablet Weight 112 - 137 mg