Flame-retardant rain jacket, high visibility, Reaven 9462
Supplier: Sioen
Sio-Start® FR
113-7332EA 276 EUR
113-7332 113-7331 113-7330 113-7329 113-7328 113-7327
Flame-retardant rain jacket, high visibility, Reaven 9462
Workwear
This waterproof and windproof jacket is made of anti-static Sio-Start FR AST fabric: 100% polyester fabric with 100% FR PU coating + AST. Besides that, it features a fixed flame-retardant cotton lining and a detachable flame-retardant body fleece lining. The outer fabric is water-repellent, while the coating on the inside attracts moisture and keeps the skin dry.
  • High tear resistance, taped seams
  • Straight collar with foldaway hood
  • Zip closure under flap with touch and close fastening
  • Two patched pockets, Napoleon pocket
  • Sleeve narrowing by press studs, elastic drawstring in hem
Back length: 85 cm
Weight: 250 g/m²
EN 531 (A B2 C1), EN 13034 (Type PB/6), EN 14116 (inside index 3; outside index 1), EN 1149-5, EN 343 (3-3), EN 471 (3-2)
Detachable lining only: EN 14058 (1XXXX)
Certifications: EN 531 (A B2 C1), EN 13034 (Type PB/6), EN 14116 (inside index 3; outside index 1), EN 1149-5, EN 343 (3-3), EN 471 (3-2)
Detachable lining only: EN 14058 (1XXXX)