You Searched For: Cameras
44  results were found
SearchResultCount:"44"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (630-2958)
Supplier: VWR Collection
Description: High performance camera with advanced software for different observation methods.
UOM: 1 * 1 items
Catalog Number: (630-2992)
Supplier: VWR Collection
Description: This high resolution C-mount camera is equipped with a 20 MP colour CMOS 1" sensor. The large sensor and cooling system make it ideal for low light ap...
UOM: 1 * 1 items
Catalog Number: (630-2661)
Supplier: VWR Collection
Description: This system is composed of a tablet and a wireless 5 MP camera. It can be attached to all microscopes with a C-mount.
UOM: 1 * 1 items
Supplier: VWR Collection
Description: These cameras provide high resolution images for everyday microscopy.
Catalog Number: (630-2957)
Supplier: VWR Collection
Description: VisiCam B5 is a C-mount and eyepiece microscope camera.
UOM: 1 * 1 items
Catalog Number: (630-1120)
Supplier: VWR Collection
Description: Analog camera that can be connected to video projectors, external monitors or a computer via the optional USB set.
UOM: 1 * 1 items
Supplier: VWR Collection
Description: High speed cameras with excellent colour reproduction for routine applications.
Catalog Number: (630-2678)
Supplier: VWR Collection
Description: This Windows® tablet PC with large LCD touch screen and integrated 5,1 MP camera provides an advanced solution for digital microscopy. For use with an...
UOM: 1 * 1 items
Catalog Number: (630-3271)
Supplier: VWR Collection
Description: Wi-Fi camera with good resolution (up to 5 MP), CMOS sensor and Wi-Fi connection, recommended for basic/general applications in education and home use...
UOM: 1 * 1 items
Supplier: LEICA MICROSYSTEMS
Description: With Flexacam microscope cameras, you can see the fine sample details you need to see with precise colors and high resolution.
New Product
Catalog Number: (TEST0560.8651)
Supplier: Testo
Description: The testo 865s thermal imager offers professional technology and concentrates on functions which are needed in a contractor's daily work. The user-fri...
UOM: 1 * 1 items
Supplier: LEICA MICROSYSTEMS
Description: Leica (brightfield) colour cameras provide outstanding colour fidelity due to state-of-the-art colour interpolation algorithms performed in the camera...
Catalog Number: (611-2257)
Supplier: VWR Collection
Description: This USB camera fits into the eyepiece tube of biological binocular and monocular microscopes. This enables users to digitise images even when the mic...
UOM: 1 * 1 items
Catalog Number: (TEST0560.8716)
Supplier: Testo
Description: Testo 871s thermal imager together with our experienced professionals to enable you to work quickly and easily with professional thermal imaging techn...
UOM: 1 * 1 items
Supplier: Testo
Description: The testo 883 thermal imager provides outstanding image quality, automatic allocation/archiving of thermal images to the measurement object and profes...
Catalog Number: (TEST0560.8684)
Supplier: Testo
Description: The testo 868s thermal imager, which makes work quicker and easier without foregoing professional thermal imaging technology. Generates error-free and...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 44
no targeter for Bottom