Blue light LED transilluminators, VWR® Blue
VWR®
730-1390EA 0 EUR
VWRI730-1390 VWRI730-1389
Blue light LED transilluminators, VWR® Blue
Blue light transilluminators are often used as an alternative to UV transilluminators when users wish to use ‘safe dyes’ instead of ethidium bromide. A blue light transilluminator is a ‘safe’ light source, in that the user is not exposed to harmful UV radiation and samples are free from photonicking.