DNA clean up, Mag-Bind® NGS Clean-IT 96 Kit
Supplier: Omega Bio-Tek
OMEGM6481-00
DNA clean up, Mag-Bind® NGS Clean-IT 96 Kit
The library construction process for next generation sequencing (NGS) requires fragment removal of smaller fragments and adaptors regardless of the sequencer platform. Obtaining a high recovery rate and a precise DNA size distribution range is key to reduce sequencing bias. The Mag-Bind® NGS Clean-IT 96 Kit is optimised for use with the library construction workflows for all the common NGS platforms, including Illumina HiSeq/GA/MiSeq, Roche 454 Genome Sequencer and Life Technologies SOLiD. The Mag-Bind® NGS Clean-IT 96 Kit is a convenient system for fast and reliable recovery of DNA fragments from various enzymatic reaction steps in the NGS library construction. The Mag-Bind® NGS Clean-IT 96 Kit can selectively and precisely recover DNA fragments free of oligonucleotides, nucleotides, and polymerase with recovery rates >50%. Users can easily select the cut-off size between 100 - 300 bp.
  • DNA recovery from enzymatic reactions can be achieved in less than 20 minutes
  • Customisable protocol to fine-tune DNA recovery size range
  • Optimised buffers that guarantee pure DNA