Safety spectacles, Voodoo
Supplier: Bollé Safety
Voodoo
111-1028EA 0 EUR
111-1028 111-1029
Safety spectacles, Voodoo
Safety Glasses
Ultra-wrap and sporty, this model perfectly combines comfort, protection and design. Available in smoke, brown and polarised brown.
  • Side protection
  • Non slip bridge
  • Anti-scratch, anti-fog coatings
  • Non slip temples
Weight: 29 g
Certifications: EN 166, EN 172