CellCultureGuard 100X
Supplier: PanReac AppliChem
A8906.0050EA
331
EUR
A8906.0050
CellCultureGuard 100X
CellCultureGuard
CellCulture Guard is a 100X concentrated solution containing a combination of novel antibiotics to protect cell cultures from contamination by microorganisms. The antibiotics in CellCultureGuard replace conventional antibiotics such as penicillin, streptomycin, nystatin and amphotericin B.