Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (13)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
Chromatography Paper
SearchPresentationType-VERTICAL
SearchResultCount:"7"
  List View Easy View (new)
Sort by:
Supplier:  GE Healthcare - Whatman
Description:   Whatman chromatography papers are widely used across the world, reflecting their purity, high quality and consistency. They are made from specially se...
Catalog Number: (WHAT3668-915)
Supplier:  GE Healthcare - Whatman
Description:   These ion exchange papers combine cellulose and large pore silica gel for separations in which both partition and adsorption are important, including ...
UOM:  1 * 25 items
Supplier:  Ahlstrom Munksjö
Description:   These chromatography filter papers are made of pure cotton linters with an alpha cellulose content of nearly 100%.
Supplier:  MACHEREY-NAGEL
Description:   Paper chromatography is the chromatographic technique separation due to partition of the analytes between special paper grades and the mobile phase, w...
Supplier:  MACHEREY-NAGEL
Description:   Filter papers with ion exchange resin.
Supplier:  Sartorius
Description:   Chromatography papers are not only ideal for chromatography, but also for routine and repetitive separations, routine analysis of proteins in serum (e...
Supplier:  VARIAN
Description:   Chromatography paper is used in thin layer chromatography applications such as evaluating radioisotope purity, QC testing.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 7  of 7
A new look and a renewed commitment
Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.
Learn More Start Shopping