You Searched For: SPE Sorbents
19  results were found
SearchResultCount:"19"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (7061-00)
Supplier: Avantor
Description: SPE sorbent for clean-up columns. Suitable for the determination of Hydrocarbon Oil Index according ISO 9377-2 and NEN 5733.
UOM: 1 * 100 g
Supplier: G-Biosciences
Description: These cross-linking agents contain at least two reactive groups that are reactive towards numerous groups, including sulphydryls, amines and carbohydr...
Supplier: MACHEREY-NAGEL
Description: The high purity CHROMABOND® adsorbents can be used for a self-filling of SPE columns or further analytical applications.
Supplier: VARIAN
Description: PL-Wang resin, 1% DVB, 0,3 mmol/g, 75 - 150 μm, 1 kg
Supplier: VARIAN
Description: Bondesil™ SCX bulk sorbent, 40 µm
Supplier: VARIAN
Description: Aminomethylstyrene resin is a particularly versatile material suitable for the attachment of a variety of spacers, handles and linkers (for use in sol...
Supplier: VARIAN
Description: This macroporous polystyrene product has a much higher loading compared to conventional functionalised silica SPE media and is compatible with all pol...
Supplier: VARIAN
Description: PL-CMS resin, 1% DVB, 0,4 mmol/g, 75 - 150 μm, 1 kg
Supplier: VARIAN
Description: PL Cl-Trt-Cl resin, 1% DVB, 1,4 mmol/g, 75 - 150 µm, 100 g
Supplier: VARIAN
Description: PL-Guanidine MP SPE, flangeless, 500 mg/6 ml
Supplier: VARIAN
Description: Strongly basic anion exchanger, Cl⁻ form. Moist, light yellow spherules.
Catalog Number: (REST26033)
Supplier: Restek
Description: Diatomaceous earth is used as a filter aid to improve extraction efficiency of densely packed soils, such as clays. By mixing the sample with diatomac...
UOM: 1 * 1 items
Supplier: VARIAN
Description: Hydromatrix™ bulk material
Catalog Number: (REST26137)
Supplier: Restek
Description: Ottawa sand is listed in several U.S. EPA methods as the specified medium for matrix spike and laboratory control blanks.
UOM: 1 * 1 items
Supplier: VARIAN
Description: PL-Rink resin, 1% DVB, 0,3 mmol/g, 75 - 150 μm, 1 kg
Catalog Number: (REST26204)
Supplier: Restek
Description: Anhydrous sodium sulphate is the most common drying agent used to remove moisture from sample extracts. Activate by heating to 400 °C for four hours b...
UOM: 1 * 5 kg
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 19
no targeter for Bottom