Barcode reader, 2D, visionmate® SR (single rack)
Supplier: Thermo Scientific
479-1250EA 5539 EUR
MATR3115
Barcode reader, 2D, visionmate® SR (single rack)
This compact, user-friendly reader decodes a full rack of 2D barcoded storage tubes in under 10 seconds.
  • Versatile: Controlled via on-screen software interface or remotely via TCP/IP connection
  • Can be connected to a computer via a single USB port
  • Integrates easily with LIMS, existing databases or tracking systems
  • Ssmall footprint and compact profile saves valuable laboratory space