Would you like to log in again?
Mats and Flooring for Cleanrooms
Dust- and particle-collecting mats and special flooring to prevent contamination in cleanrooms.
Mats and Flooring for Cleanrooms Particles have a basic tendency to settle on flat surfaces, due to the varying adhesion factors throughout the cleanrooms and strongly influenced by the airflow characteristics. The flooring is the final location where particles settle or remain as captured contamination.
Adhesive mats, permanent mats and special floor coverings are essential to control particle contamination in front of the cleanroom. For optimal effective control of contamination in cleanrooms and controlled environments, the combination of permanent mats, special flooring and adhesive mats is highly recommended.