Antifatigue Cleanroom Mats
Antifatigue cleanroom mats have the comfort of polyurethane with the added benefit of being easy to clean due to the closed cell / non porous top surface. Constructed of virgin raw materials, the mats are solid throughout. They feature extremely good chemical (except for organic solvents) and UV resistance, as well as anti-static properties surface.
All mats can be customized to any size or shape and have 12 year warranty.
Product Selection
Antifatigue Mats
Please choose from a list of products and refine your search by attributes...