Solid Phase Extraction
Avantor offers a variety of J.T.Baker® brand silica- and polymer-based BAKERBOND spe columns, high performance J.T.Baker® BAKERBOND Speedisk columns and disks, and J.T.Baker® standard vacuum processors to improve and simplify sample clean-up and concentration.
  • J.T.Baker® BAKERBOND spe columns are recommended when standard performance with good economy is needed
  • J.T.Baker® BAKERBOND Speedisk columns are recommended when higher levels of speed and performance are required
Request your application note
Product Selection
J.T.Baker® BAKERBOND spe™ extraction disks
50mm disks for samples from 200ml to 2L. These are not cardridge or membrane disks.
J.T.Baker® BAKERBOND spe™ extraction columns
1ml, 3ml and 6ml columns, round-rimmed and earshaped in ultraclean polypropylene or glass
J.T.Baker® BAKERBOND sorbents for SPE
Sorbent bulk packing material
J.T.Baker® general SPE accessories
General SPE accessories such as reservoirs and adaptors