Would you like to log in again?
ACE® HPLC and UHPLC Columns from ACE® Ltd
Advanced Chromatography Technologies manufacture the ultra-inert ACE® UHPLC/HPLC column range. ACE® phases incorporate the latest developments in LC stationary phase design, providing chromatographers with more choices for alternative selectivity, without compromising stability or robustness.
ACE® LC phases have been specifically designed to help solve chromatographic challenges and include the unique C18-AR, C18-PFP, C18-Amide, CN-ES, SuperC18 and SuperPhenylHexyl phases, as well as a wide range of other 100 Å and 300 Å phases. Discover how ACE® selectivities can help you with your chromatographic challenges. ACE® - superior LC columns.
Download the ACE®complete application guide Contact technical support