Microgard® protective clothing - featuring 2000Ts Plus
Product of choice for pharmaceutical workers around the world!
MICROGARD® 2000 Ts Plus is designed to allow water vapour (perspiration) to escape from the suit yet will withstand saturation of liquid chemicals and filter 100% of particulates down to 0,01 microns in size*. (*EMSL test method).
- Anti-static (according to EN 1149-5)
- Low linting: Reduced risk of contamination
- Silicone-free: Critical in spray painting applications
- Comfort: Moisture vapour permeable to help reduce the risk of heat stress
- Thumb loops help prevent sleeve movement when working above your head
- Chin strap helps to reduce risk of cross-contamination