Cleanroom Sponges
Sponges are ideal for the removal of strong and pasty contamination. They are available autoclavable and with material resistant to strong disinfectants. For cleaning cleanroom curtains and softwalls, laminar flow benches and isolators. Also suitable as an intermediate layer for transport of critical products.
In a variety of sizes (also customized sizes available) and in different packaging units available.
Product Selection
Sponges
Please choose from a list of products and refine your search by attributes...