Chemicals
VWR are proud to offer one of the largest selection of chemicals from one source to our customers. As well as extensive investment in our own chemical production , our mission is to make getting the products you need effortless.
Chemicals by method
Chemicals for sample Preparation
Sample preparation is as important as the analysis of your sample. Make sure you use the right reagents to avoid contamination of your sample.
HPLC
The right choice of chemical is crucial for your chromatography process. Below an overview of our chemical range for chromatography.
GC
In gas chromatography, sample preparation is critical to obtain the right result. Choosing the right chemicals for your method and application will help you for a succesful analysis.
Titration
Everything you need for this common quantitative technique from classical glassware to automatic titrators, including reagents and accessories.
Water determination
Choice of compact volumetric or coulometric Karl Fisher titrators and related reagents for water determination.
pH measurement
pH measurements using a pH meter can only be correct and precise when you are using the right buffer for your instrument.
Conductivity measurement
Conductivity measurement of an aqueous solution is a method used to determine amount of nutrients, salts and impurities. Conductivity standards are crucial to calibrate your conductivity meter.
Physical property standards
Many different ranges for physical measurements: Viscosity standards, melting point standards, density standards, refractive index and Brix standards are available.
UV/VIS Spectroscopy
UV/Vis analysis with a spectrophotometer can require different types of chemicals. First, a specific grade of solvents is developed to avoid optical impurities of your sample. Second, the spectrophotometer...
NMR (Nuclear Magnetic Resonance)
NMR (Nuclear Magnetic Resonance) is used as a standard analysis method for structural determination of an organic compound.
AAS/ICP
In metal analysis, elemental evidence is now gathered from the tiniest traces in the ppm, ppb and ppt ranges with sophisticated instrumentation like AAS, ICP-OES or, more recently, ICP-MS.
Organic synthesis
If you’re creating new molecules in your pharmaceutical or chemical research or university laboratory, VWR offers an extensive range of products to help you with your discoveries.
Nitrogen determination
The Kjeldahl method is used to determine the nitrogen content of a sample. In this analysis, the sample is digested with a strong acid and the free nitrogen reacts to form ammonium sulfate.
Dissolution testing
Dissolution media preparation can be a time consuming task when done manually. J.T.Baker® DILUT-IT™ dissolution media concentrates are pre-mixed concentrates that significantly reduce...
Custom solutions
VWR enables the advancement of science by providing high-quality chemicals and services, customized to your product or manufacturing needs.
General lab supplies
Some products are essential in every lab. Improve safety in your lab with VWRC, etc.
Chemical manager
Your inventory should benefit your scientific initiative, not delay it – and if your supply management is not at its most efficient, your staff could be spending valuable time just trying to get the products they need.