Cleanroom Packaging
We offer a wide range of products for demanding packaging applications in cleanrooms and controlled environments such as suitability for sterilisation, low particle contamination, resistance to most chemicals, high quality sealing or antistatic. (Flat) bags, easy-tear-bags or tubes are available. Transparent foils offer the advantage of product visibility. The range also includes foils suitable for printing.
All cleanroom packaging types are available for various cleanroom classes as well as packages for machines and devices of all sizes or other oversized sterilised goods. Sealing devices are available upon request.
Tyvek Packaging
Made of transparent foil with very low gas residue, extremely low particle contamination and outstanding barrier characteristics. Available in various cleanroom classes.
Transparent Paper Packaging
Transparent and printable foil. Unlimited suitability for steam-, ethylene-oxide- and formaldehyde-sterilisation. As flat bag or tube.
Polyethylene Packaging
Ultra-clean, clear polyethylene. Suitable for gamma sterilisation and resistant to most chemicals. Flat bags, tubes and packages for machines and devices of all sizes are available.
Antistatic Polyethylene Packaging
Made from ultra-clean polyethylene, non-corrosive, resistant to most chemicals and easily heat-sealed.
Knives and Cutters
For easy opening of packaging and other cutting work in cleanrooms. Reusable and disposable versions available.